missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKCE Protein
A cDNA sequence encoding the PRKCE was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13145-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
PRKCE Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MVVFNGLLKI KICEAVSLKP TAWSLRHAVG PRPQTFLLDP YIALNVDDSR IGQTATKQKT NSPAWHDEFV TDVCNGRKIE LAVFHDAPIG YDDFVANCTI QFEELLQNGS RHFEDWIDLE PEGRVYVIID LSGSSGEAPK DNEERVFRER MRPRKRQGAV RRRVHQVNGH KFMATYLRQP TYCSHCRDFI WGVIGKQGYQ CQVCTCVVHK RCHELIITKC AGLKKQETPD QVGSQRFSVN MPHKFGIHNY KVPTFCDHCG SLLWGLLRQG LQCKVCKMNV HRRCETNVAP NCGVDARGIA KVLADLGVTP DKITNSGQRR KKLIAGAESP QPASGSSPSE EDRSKSAPTS PCDQEIKELE NNIRKALSFD NRGEEHRAAS SPDGQLMSPG ENGEVRQGQA KRLGLDEFNF IKVLGKGSFG KVMLAELKGK DEVYAVKVLK KDVILQDDDV DCTMTEKRIL ALARKHPYLT QLYCCFQTKD RLFFVMEYVN GGDLMFQIQR SRKFDEPRSR FYAAEVTSAL MFLHQHGVIY RDLKLDNILL DAEGHCKLAD FGMCKEGILN GVTTTTFCGT PDYIAPEILQ ELEYGPSVDW WALGVLMYEM MAGQPPFEAD NEDDLFESIL HDDVLYPVWL SKEAVSILKA FMTKNPHKRL GCVASQNGED AIKQHPFFKE IDWVLLEQKK IKPPFKPRIK TKRDVNNFDQ DFTREEPVLT LVDEAIVKQI NQEEFKGFSY FGEDLMP | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRKCE is supplied in 20mM Tris, pH 7.4, 0.1M NaCl, 20% glycerol, 1mM DTT, 0.1mM EDTA, 0.2mM PMSF, 1mM benzamidine and 0.03% Brij-35. |