missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRKAR2A Protein

A cDNA sequence encoding the PRKAR2A was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13144-5ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15962639

Menge Preis
1 128.22 CHF / 5 Mikrogramm
10 117.77 CHF / 5 Mikrogramm
Voraussichtliches Versanddatum: 03-11-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

PRKAR2A Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PRKAR2A protein solution (0.5 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSHIQIP PGLTELLQGY TVEVLRQQPP DLVEFAVEYF TRLREARAPA SVLPAATPRQ SLGHPPPEPG PDRVADAKGD SESEEDEDLE VPVPSRFNRR VSVCAETYNP DEEEEDTDPR VIHPKTDEQR CRLQEACKDI LLFKNLDQEQ LSQVLDAMFE RIVKADEHVI DQGDDGDNFY VIERGTYDIL VTKDNQTRSV GQYDNRGSFG ELALMYNTPR AATIVATSEG SLWGLDRVTF RRIIVKNNAK KRKMFESFIE SVPLLKSLEV SERMKIVDVI GEKIYKDGER IITQGEKADS FYIIESGEVS ILIRSRTKSN KDGGNQEVEI ARCHKGQYFG ELALVTNKPR AASAYAVGDV KCLVMDVQAF ERLLGPCMDI MKRNISHYEE QLVKMFGSSV DLGNLGQ
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate