missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAR2A Protein
A cDNA sequence encoding the PRKAR2A was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | PRKAR2A Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15962639
|
enQuireBio™
QP13144-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15952639
|
enQuireBio™
QP13144-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15942639
|
enQuireBio™
QP13144-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PRKAR2A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRKAR2A protein solution (0.5 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSHIQIP PGLTELLQGY TVEVLRQQPP DLVEFAVEYF TRLREARAPA SVLPAATPRQ SLGHPPPEPG PDRVADAKGD SESEEDEDLE VPVPSRFNRR VSVCAETYNP DEEEEDTDPR VIHPKTDEQR CRLQEACKDI LLFKNLDQEQ LSQVLDAMFE RIVKADEHVI DQGDDGDNFY VIERGTYDIL VTKDNQTRSV GQYDNRGSFG ELALMYNTPR AATIVATSEG SLWGLDRVTF RRIIVKNNAK KRKMFESFIE SVPLLKSLEV SERMKIVDVI GEKIYKDGER IITQGEKADS FYIIESGEVS ILIRSRTKSN KDGGNQEVEI ARCHKGQYFG ELALVTNKPR AASAYAVGDV KCLVMDVQAF ERLLGPCMDI MKRNISHYEE QLVKMFGSSV DLGNLGQ | |
Greater than 85.0% as determined by SDS-PAGE. |