missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPM1F Protein
A cDNA sequence encoding the PPM1F was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13119-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
PPM1F Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSGAPQ KSSPMASGAE ETPGFLDTLL QDFPALLNPE DPLPWKAPGT VLSQEEVEGE LAELAMGFLG SRKAPPPLAA ALAHEAVSQL LQTDLSEFRK LPREEEEEEE DDDEEEKAPV TLLDAQSLAQ SFFNRLWEVA GQWQKQVPLA ARASQRQWLV SIHAIRNTRR KMEDRHVSLP SFNQLFGLSD PVNRAYFAVF DGHGGVDAAR YAAVHVHTNA ARQPELPTDP EGALREAFRR TDQMFLRKAK RERLQSGTTG VCALIAGATL HVAWLGDSQV ILVQQGQVVK LMEPHRPERQ DEKARIEALG GFVSHMDCWR VNGTLAVSRA IGDVFQKPYV SGEADAASRA LTGSEDYLLL ACDGFFDVVP HQEVVGLVQS HLTRQQGSGL RVAEELVAAA RERGSHDNIT VMVVFLRDPQ ELLEGGNQGE GDPQAEGRRQ DLPSSLPEPE TQAPPRS | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PPM1F solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |