missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLR2J2 Protein

A cDNA sequence encoding the POLR2J2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13103-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15912519

  • 7190.00 CHF / 1 Milligramm


Verfügbar ab
05-05-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

POLR2J2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The POLR2J2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol, 1mM DTT and 250mM Imidazole.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMNAPPAF ESFLLFEGEK ITINKDTKVP KACLFTINKE DHTLGNIIKS QLLKDPQVLF AGYKVPHPLE HKIIIRVQTT PDYSPQEAFT NAITDLISEL SLLEERFRTC LLPLRLLP
Videos
Dokumentation