missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2J Protein
A cDNA sequence encoding the POLR2J was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13102-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
POLR2J Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
POLR2J protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 5mM DTT and 2mM EDTA. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNAPPAF ESFLLFEGEK KITINKDTKV PNACLFTINK EDHTLGNIIK SQLLKDPQVL FAGYKVPHPL EHKIIIRVQT TPDYSPQEAF TNAITDLISE LSLLEERFRV AIKDKQEGIE |