missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2F Protein
A cDNA sequence encoding the POLR2F was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13099-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
POLR2F Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSDNEDN FDGDDFDDVE EDEGLDDLEN AEEEGQENVE ILPSGERPQA NQKRITTPYM TKYERARVLG TRALQIAMCA PVMVELEGET DPLLIAMKEL KARKIPIIIR RYLPDGSYED WGVDELIITD | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLR2F solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |