missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR1C Protein
A cDNA sequence encoding the POLR1C was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13095-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
POLR1C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAASQAV EEMRSRVVLG EFGVRNVHTT DFPGNYSGYD DAWDQDRFEK NFRVDVVHMD ENSLEFDMVG IDAAIANAFR RILLAEVPTM AVEKVLVYNN TSIVQDEILA HRLGLIPIHA DPRLFEYRNQ GDEEGTEIDT LQFRLQVRCT RNPHAAKDSS DPNELYVNHK VYTRHMTWIP LGNQADLFPE GTIRPVHDDI LIAQLRPGQE IDLLMHCVKG IGKDHAKFSP VATASYRLLP DITLLEPVEG EAAEELSRCF SPGVIEVQEV QGKKVARVAN PRLDTFSREI FRNEKLKKVV RLARVRDHYI FSVESTGVLP PDVLVSEAIK VLMGKCRRFL DELDAVQMD | |
Greater than 80.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
POLR1C protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |