missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PNPO Protein

A cDNA sequence encoding the PNPO was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13085-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15962459

  • 3905.00 CHF / 1 Milligramm
Verfügbar ab: 28-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

PNPO Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDPVKQFAAW FEEAVQCPDI GEANAMCLAT CTRDGKPSAR MLLLKGFGKD GFRFFTNFES RKGKELDSNP FASLVFYWEP LNRQVRVEGP VKKLPEEEAE CYFHSRPKSS QIGAVVSHQS SVIPDREYLR KKNEELEQLY QDQEVPKPKS WGGYVLYPQV MEFWQGQTNR LHDRIVFRRG LPTGDSPLGP MTHRGEEDWL YERLAP
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PNPO solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 0.1mM PMSF.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote