missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLAP Protein
A cDNA sequence encoding the PLAP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13076-2ug
Additional Details : Gewicht : 0.01000kg
Specifications
250 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MLGPCMLLLL LLLGLRLQLS LGIIPVEEEN PDFWNREAAE ALGAAKKLQP AQTAAKNLII FLGDGMGVST VTAARILKGQ KKDKLGPEIP LAMDRFPYVA LSKTYNVDKH VPDSGATATA YLCGVKGNFQ TIGLSAAARF NQCNTTRGNE VISVMNRAKK AGKSVGVVTT TRVQHASPAG TYAHTVNRNW YSDADVPASA RQEGCQDIAT QLISNMDIDV ILGGGRKYMF RMGTPDPEYP DDYSQGGTRL DGKNLVQEWL AKRQGARYVW NRTELMQASL DPSVTHLMGL FEPGDMKYEI HRDSTLDPSL MEMTEAALRL LSRNPRGFFL FVEGGRIDHG HHESRAYRAL TETIMFDDAI ERAGQLTSEE DTLSLVTADH SHVFSFGGYP LRGSSIFGLA PGKARDRKAY TVLLYGNGPG YVLKDGARPD VTESESGSPE YRQQSAVPLD EETHAGEDVA VFARGPQAHL VHGVQEQTFI AHVMAFAACL EPYTACDLAP PAGTTDAAHP GRSVVPALLP LLAGTLLLLE TATAP |
PLAP Protein | |
Research Use Only | |
ALPP | |
Human | |
GST | |
PLAP in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |