missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA2G10 Protein
A cDNA sequence encoding the PLA2G10 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13071-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PLA2G10 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGPLPVCLPI MLLLLLPSLL LLLLLPGPGS GEASRILRVH RRGILELAGT VGCVGPRTPI AYMKYGCFCG LGGHGQPRDA IDWCCHGHDC CYTRAEEAGC SPKTERYSWQ CVNQSVLCGP AENKCQELLC KCDQEIANCL AQTEYNLKYL FYPQFLCEPD SPKCD | |
Greater than 95% as determined by SDS PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Sterile filtered and lyophilized from 0.5 mg/ml in 0.01M Tris buffer pH 7.2. |