missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA1A Protein
A cDNA sequence encoding the PLA1A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13070-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
51365 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDAPPTPQ PKCADFQSAN LFEGTDLKVQ FLLFVPSNPS CGQLVEGSSD LQNSGFNATL GTKLIIHGFR VLGTKPSWID TFIRTLLRAT NANVIAVDWI YGSTGVYFSA VKNVIKLSLE ISLFLNKLLV LGVSESSIHI IGVSLGAHVG GMVGQLFGGQ LGQITGLDPA GPEYTRASVE ERLDAGDALF VEAIHTDTDN LGIRIPVGHV DYFVNGGQDQ PGCPTFFYAG YSYLICDHMR AVHLYISALE NSCPLMAFPC ASYKAFLAGR CLDCFNPFLL SCPRIGLVEQ GGVKIEPLPK EVKVYLLTTS SAPYCMHHSL VEFHLKELRN KDTNIEVTFL SSNITSSSKI TIPKQQRYGK GIIAHATPQC QINQVKFKFQ SSNRVWKKDR TTIIGKFCTA LLPVNDREKM VCLPEPVNLQ ASVTVSCDLK IACV | |
Greater than 80% as determined by SDS-PAGE. |
PLA1A Protein | |
Research Use Only | |
PLA1A | |
Human | |
His | |
The PLA1A solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |