missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PITPNA Protein
A cDNA sequence encoding the PITPNA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13060-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
PITPNA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVLLKEYRVI LPVSVDEYQV GQLYSVAEAS KNETGGGEGV EVLVNEPYEK DGEKGQYTHK IYHLQSKVPT FVRMLAPEGA LNIHEKAWNA YPYCRTVITN EYMKEDFLIK IETWHKPDLG TQENVHKLEP EAWKHVEAVY IDIADRSQVL SKDYKAEEDP AKFKSIKTGR GPLGPNWKQE LVNQKDCPYM CAYKLVTVKF KWWGLQNKVE NFIHKQERRL FTNFHRQLFC WLDKWVDLTM DDIRRMEEET KRQLDEMRQK DPVKGMTADD | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PITPNA solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol, 1mM EDTA. |