missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PITPNA Protein
A cDNA sequence encoding the PITPNA was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | PITPNA Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15902389
|
enQuireBio™
QP13060-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15992379
|
enQuireBio™
QP13060-25UG |
25 μg |
203.00 CHF
25 Mikrogramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982379
|
enQuireBio™
QP13060-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spécification
PITPNA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PITPNA solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol, 1mM EDTA. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVLLKEYRVI LPVSVDEYQV GQLYSVAEAS KNETGGGEGV EVLVNEPYEK DGEKGQYTHK IYHLQSKVPT FVRMLAPEGA LNIHEKAWNA YPYCRTVITN EYMKEDFLIK IETWHKPDLG TQENVHKLEP EAWKHVEAVY IDIADRSQVL SKDYKAEEDP AKFKSIKTGR GPLGPNWKQE LVNQKDCPYM CAYKLVTVKF KWWGLQNKVE NFIHKQERRL FTNFHRQLFC WLDKWVDLTM DDIRRMEEET KRQLDEMRQK DPVKGMTADD | |
Greater than 95.0% as determined by SDS-PAGE. |