missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PINK1 Protein
A cDNA sequence encoding the PINK1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | PINK1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15982369
|
enQuireBio™
QP13056-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972369
|
enQuireBio™
QP13056-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15962369
|
enQuireBio™
QP13056-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PINK1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The PINK1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 5% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MYLIGQSIGK GCSAAVYEAT MPTLPQNLEV TKSTGLLPGR GPGTSAPGEG QERAPGAPAF PLAIKMMWNI SAGSSSEAIL NTMSQELVPA SRVALAGEYG AVTYRKSKRG PKQLAPHPNI IRVLRAFTSS VPLLPGALVD YPDVLPSRLH PEGLGHGRTL FLVMKNYPCT LRQYLCVNTP SPRLAAMMLL QLLEGVDHLV QQGIAHRDLK SDNILVELDP DGCPWLVIAD FGCCLADESI GLQLPFSSWY VDRGGNGCLM APEVSTARPG PRAVIDYSKA DAWAVGAIAY EIFGLVNPFY GQGKAHLESR SYQEAQLPAL PESVPPDVRQ LVRALLQREA SKRPSARVAA NVL | |
Greater than 90% as determined by SDS-PAGE. |