missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGRN Protein
A cDNA sequence encoding the PGRN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10817-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
2896 | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Activates ERK phosphorylation in THP-1 and MCF10A cells and induces cell migration of THP-1 monocytic leukemia cells. | |
Human | |
Untagged | |
The protein solution contains 1xPBS. |
PGRN Protein | |
Research Use Only | |
GRN | |
Recombinant Protein | |
Human (HEK293) | |
MWTLVSWVAL TAGLVAGTRC PDGQFCPVAC CLDPGGASYS CCRPLLDKWP TTLSRHLGGP CQVDAHCSAG HSCIFTVSGT SSCCPFPEAV ACGDGHHCCP RGFHCSADGR SCFQRSGNNS VGAIQCPDSQ FECPDFSTCC VMVDGSWGCC PMPQASCCED RVHCCPHGAF CDLVHTRCIT PTGTHPLAKK LPAQRTNRAV ALSSSVMCPD ARSRCPDGST CCELPSGKYG CCPMPNATCC SDHLHCCPQD TVCDLIQSKC LSKENATTDL LTKLPAHTVG DVKCDMEVSC PDGYTCCRLQ SGAWGCCPFT QAVCCEDHIH CCPAGFTCDT QKGTCEQGPH QVPWMEKAPA HLSLPDPQAL KRDVPCDNVS SCPSSDTCCQ LTSGEWGCCP IPEAVCCSDH QHCCPQGYTC VAEGQCQRGS EIVAGLEKMP ARRASLSHPR DIGCDQHTSC PVGQTCCPSL GGSWACCQLP HAVCCEDRQH CCPAGYTCNV KARSCEKEVV SAQPATFLAR SPHVGVKDVE CGEGHFCHDN QTCCRDNRQG WACCPYRQGV CCADRRHCCP AGFRCAARGT KCLRREAPRW DAPLRDPALR QLL | |
Greater than 95.0% as determined by SDS-PAGE. |