missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PGLYRP1 Protein

A cDNA sequence encoding the PGLYRP1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13033-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982299

  • 188.00 CHF / 10 Mikrogramm


Verfügbar ab
29-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

PGLYRP1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PGLYRP1 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.0.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASQETEDPACCS PIVPRNEWKA LASECAQHLS LPLRYVVVSH TAGSSCNTPA SCQQQARNVQ HYHMKTLGWC DVGYNFLIGE DGLVYEGRGW NFTGAHSGHL WNPMSIGISF MGNYMDRVPT PQAIRAAQGL LACGVAQGAL RSNYVLKGHR DVQRTLSPGN QLYHLIQNWP HYRSP
Videos
Dokumentation