missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human pGH 22kDa Protein
A cDNA sequence encoding the pGH 22kDa was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13030-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
pGH 22kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAAGSRTSLL LAFGLLCLSW LQEGSAFPTI PLSRLFDNAM LRARRLYQLA YDTYQEFEEA YILKEQKYSF LQNPQTSLCF SESIPTPSNR VKTQQKSNLE LLRISLLLIQ SWLEPVQLLR SVFANSLVYG ASDSNVYRHL KDLEEGIQTL MWRLEDGSPR TGQIFNQSYS KFDTKSHNDD ALLKNYGLLY CFRKDMDKVE TFLRIVQCRS VEGSCGF | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3 previously adjusted pH 8-9. |