missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGAM2 Protein
A cDNA sequence encoding the PGAM2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 203.00 CHF
Spezifikation
Name | PGAM2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992279
|
enQuireBio™
QP13026-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982279
|
enQuireBio™
QP13026-25UG |
25 μg |
203.00 CHF
25 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PGAM2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PGAM2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MATHRLVMVR HGESTWNQEN RFCGWFDAEL SEKGTEEAKR GAKAIKDAKM EFDICYTSVL KRAIRTLWAI LDGTDQMWLP VVRTWRLNER HYGGLTGLNK AETAAKHGEE QVKIWRRSFD IPPPPMDEKH PYYNSISKER RYAGLKPGEL PTCESLKDTI ARALPFWNEE IVPQIKAGKR VLIAAHGNSL RGIVKHLEGM SDQAIMELNL PTGIPIVYEL NKELKPTKPM QFLGDEETVR KAMEAVAAQG KAK | |
Greater than 95.0% as determined by SDS-PAGE. |