missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PFN2 Protein

A cDNA sequence encoding the PFN2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13021-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15962269

  • 7354.84 CHF / 1 Milligramm


Voraussichtliches Versanddatum: 10-11-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

PFN2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PFN2 protein solution (1 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAGWQSYVDN LMCDGCCQEA AIVGYCDAKY VWAATAGGVF QSITPIEIDM IVGKDREGFF TNGLALGAKK CSVIRDSLYV DGDCTMDIRT KSQGGEPTYN VAVGRAGRVL VFVMGKEGVH GGGLNKKAYS MAKYLRDSGF
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate