missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PFDN4 Protein
A cDNA sequence encoding the PFDN4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13016-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PFDN4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAATMKKAAA EDVNVTFEDQ QKINKFARNT SRITELKEEI EVKKKQLQNL EDACDDIMLA DDDCLMIPYQ IGDVFISHSQ EETQEMLEEA KKNLQEEIDA LESRVESIQR VLADLKVQLY AKFGSNINLE ADES | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PFDN4 protein solution (1 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 7.5), 50mM NaCl, 1mM DTT and 10% glycerol. |