missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PFDN4 Protein

A cDNA sequence encoding the PFDN4 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13016-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15912259

  • 7525.00 CHF / 1 Milligramm
Verfügbar ab: 29-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

PFDN4 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAATMKKAAA EDVNVTFEDQ QKINKFARNT SRITELKEEI EVKKKQLQNL EDACDDIMLA DDDCLMIPYQ IGDVFISHSQ EETQEMLEEA KKNLQEEIDA LESRVESIQR VLADLKVQLY AKFGSNINLE ADES
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PFDN4 protein solution (1 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 7.5), 50mM NaCl, 1mM DTT and 10% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote