missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PF4V1 Protein
A cDNA sequence encoding the PF4V1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13013-10ug
Additional Details : Gewicht : 0.01000kg
Specifications
5197 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFARAEAE EDGDLQCLCV KTTSQVRPRH ITSLEVIKAG PHCPTAQLIA TLKNGRKICL DLQALLYKKI IKEHLES | |
Greater than 85.0% as determined by SDS-PAGE. |
PF4V1 Protein | |
Research Use Only | |
PF4V1 | |
Human | |
His | |
PF4V1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 1mM DTT. |