Learn More
enQuireBio™ Recombinant Human PELI2 Protein
A cDNA sequence encoding the PELI2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | PELI2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15922229
|
enQuireBio™
QP13007-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15902229
|
enQuireBio™
QP13007-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15912229
|
enQuireBio™
QP13007-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PELI2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PELI2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMFSPGQE EHCAPNKEPV KYGELVVLGY NGALPNGDRG RRKSRFALYK RPKANGVKPS TVHVISTPQA SKAISCKGQH SISYTLSRNQ TVVVEYTHDK DTDMFQVGRS TESPIDFVVT DTISGSQNTD EAQITQSTIS RFACRIVCDR NEPYTARIFA AGFDSSKNIF LGEKAAKWKN PDGHMDGLTT NGVLVMHPRG GFTEESQPGV WREISVCGDV YTLRETRSAQ QRGKLVESET NVLQDGSLID LCGATLLWRT ADGLFHTPTQ KHIEALRQEI NAARPQCPVG LNTLAFPSIN RKEVVEEKQP WAYLSCGHVH GYHNWGHRSD TEANERECPM CRTVGPYVPL WLGCEAGFYV DAGPPTHAFT PCGHVCSEKS AKYWSQIPLP HGTHAFHAAC PFCATQLVGE QNCIKLIFQG PID | |
Greater than 80.0% as determined by SDS-PAGE. |