missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PEF1 Protein
A cDNA sequence encoding the PEF1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13006-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PEF1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASY PYRQGCPGAA GQAPGAPPGSYYPGPPNSGG QYGSGLPPGG GYGGPAPGGP YGPPAGGGPY GHPNPGMFPS GTPGGPYGGAAPGGPYGQPP PSSYGAQQPG LYGQGGAPPN VDPEAYSWFQ SVDSDHSGYI SMKELKQALVNCNWSSFNDE TCLMMINMFD KTKSGRIDVY GFSALWKFIQ QWKNLFQQYD RDRSGSISYTELQQALSQMG YNLSPQFTQL LVSRYCPRSA NPAMQLDRFI QVCTQLQVLT EAFREKDTAVQGNIRLSFED FVTMTASRML | |
Greater than 85.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PEF1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50% glycerol and 0.2M NaCl. |