missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PECR Protein
A cDNA sequence encoding the PECR was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | PECR Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15972209
|
enQuireBio™
QP13004-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15962209
|
enQuireBio™
QP13004-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15952209
|
enQuireBio™
QP13004-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PECR Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PECR solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 2mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMASWAK GRSYLAPGLL QGQVAIVTGG ATGIGKAIVK ELLELGSNVV IASRKLERLK SAADELQANL PPTKQARVIP IQCNIRNEEE VNNLVKSTLD TFGKINFLVN NGGGQFLSPA EHISSKGWHA VLETNLTGTF YMCKAVYSSW MKEHGGSIVN IIVPTKAGFP LAVHSGAARA GVYNLTKSLA LEWACSGIRI NCVAPGVIYS QTAVENYGSW GQSFFEGSFQ KIPAKRIGVP EEVSSVVCFL LSPAASFITG QSVDVDGGRS LYTHSYEVPD HDNWPKGAGD LSVVKKMKET FKEKAKL | |
Greater than 95% as determined by SDS-PAGE. |