missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDPN Protein
A cDNA sequence encoding the PDPN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12999-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PDPN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASTGQPEDD TETTGLEGGV AMPGAEDDVV TPGTSEDRYK SGLTTLVATS VNSVTGIRIE DLPTSESTVH AQEQSPSATA SNVATSHSTE KVDGDTQTTV EKDGLSTVTL | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution contains 20mM Tris-HCl pH 7.5 and 0.1M NaCl. |