missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDE6D Protein
A cDNA sequence encoding the PDE6D was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12990-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
5147 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSAKDERARE ILRGFKLNWM NLRDAETGKI LWQGTEDLSV PGVEHEARVP KKILKCKAVS RELNFSSTEQ MEKFRLEQKV YFKGQCLEEW FFEFGFVIPN STNTWQSLIE AAPESQMMPA SVLTGNVIIE TKFFDDDLLV STSRVRLFYV GSHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
PDE6D Protein | |
Research Use Only | |
PDE6D | |
Human | |
His | |
The PDE6D protein solution contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 100mM NaCl. |