missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCSK1N Protein
A cDNA sequence encoding the PCSK1N was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12981-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
27344 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARPVKE PRGLSAASPP LAETGAPRRF RRSVPRGEAA GAVQELARAL AHLLEAERQE RARAEAQEAE DQQARVLAQL LRVWGAPRNS DPALGLDDDP DAPAAQLARA LLRARLDPAA LAAQLVPAPV PAAALRPRPP VYDDGPAGPD AEEAGDETPD VDPELLRYLL GRILAGSADS EGVAAPRRLR RAADHDVGSE LPPEGVLGAL LRVKRLETPA PQVPARRLLP P | |
Greater than 85.0% as determined by SDS-PAGE. |
PCSK1N Protein | |
Research Use Only | |
PCSK1N | |
Human | |
His | |
The PCSK1N solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |