missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p59-Fyn Protein
A cDNA sequence encoding the p59-Fyn was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12949-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
2534 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGCVQCKDKE ATKLTEERDG SLNQSSGYRY GTDPTPQHYP SFGVTSIPNY NNFHAAGGQG LTVFGGVNSS SHTGTLRTRG GTGVTLFVAL YDYEARTEDD LSFHKGEKFQ ILNSSEGDWW EARSLTTGET GYIPSNYVAP VDSIQAEEWY FGKLGRKDAE RQLLSFGNPR GTFLIRESET TKGAYSLSIR DWDDMKGDHV KHYKIRKLDN GGYYITTRAQ FETLQQLVQH YSERAAGLCC RLVVPCHKGM PRLTDLSVKT KDVWEIPRES LQLIKRLGNG QFGEVWMGTW NGNTKVAIKT LKPGTMSPES FLEEAQIMKK LKHDKLVQLY AVVSEEPIYI VTEYMNKGSL LDFLKDGEGR ALKLPNLVDM AAQVAAGMAY IERMNYIHRD LRSANILVGN GLICKIADFG LARLIEDNEY TARQGAKFPI KWTAPEAALY GRFTIKSDVW SFGILLTELV TKGRVPYPGM NNREVLEQVE RGYRMPCPQD CPISLHELMI HCWKKDPEER PTFEYLQSFL EDYFTATEPQ YQPGENL |
p59-Fyn Protein | |
Research Use Only | |
FYN | |
Human | |
GST | |
p59-Fyn in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |