missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p16-INK4a, TAT Protein
A cDNA sequence encoding the p16-INK4a, TAT was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Gen-ID (Entrez) | 1029 |
---|---|
Name | p16-INK4a, TAT Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | CDKN2A |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15985439
|
enQuireBio™
QP14024-EC-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965439
|
enQuireBio™
QP14024-25UG |
25 μg |
203.00 CHF
25 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975439
|
enQuireBio™
QP14024-EC-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
1029 | |
Research Use Only | |
CDKN2A | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
p16-INK4a, TAT Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
EPAAGSSMEP SADWLATAAA RGRVEEVRAL LEAGALPNAP NSYGRRPIQV MMMGSARVAE LLLLHGAEPN CADPATLTRP VHDAAREGFL DTLVVLHRAG ARLDVRDAWG RLPVDLAEEL GHRDVARYLR AAAGGTRGSN HARIDAAEGP SDIPDGGYGR KKRRQRRR | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |