missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human P Selectin Protein
A cDNA sequence encoding the P Selectin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12945-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
P Selectin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANCQIAILY QRFQRVVFGI SQLLCFSALI SELTNQKEVA AWTYHYSTKA YSWNISRKYC QNRYTDLVAI QNKNEIDYLN KVLPYYSSYY WIGIRKNNKT WTWVGTKKAL TNEAENWADN EPNNKRNNED CVEIYIKSPS APGKWNDEHC LKKKHALCYT ASCQDMSCSK QGECLETIGN YTCSCYPGFY GPECEYVREC GELELPQHVL MNCSHPLGNF SFNSQCSFHC TDGYQVNGPS KLECLASGIW TNKPPQCLAA QCPPLKIPER GNMTCLHSAK AFQHQSSCSF SCEEGFALVG PEVVQCTASG VWTAPAPVCK AVQCQHLEAP SEGTMDCVHP LTAFAYGSSC KFECQPGYRV RGLDMLRCID SGHWSAPLPT CEAISCEPLE SPVHGSMDCS PSLRAFQYDT NCSFRCAEGF MLRGADIVRC DNLGQWTAPA PVCQALQCQD LPVPNEARVN CSHPFGAFRY QSVCSFTCNE GLLLVGASVL QCLATGNWNS VPPECQAIPC TPLLSPQNGT MTCVQPLGSS SYKSTCQFIC DEGYSLSGPE RLDCTRSGRW TDSPPMCEAI KCPELFAPEQ GSLDCSDTRG EFNVGSTCHF SCDNGFKLEG PNNVECTTSG RWSATPPTCK GIASLPTPGL QCPALTTPGQ GTMYCRHHPG TFGFNTTCYF GCNAGFTLIG DSTLSCRPSG QWTAVTPACR AVKCSELHVN KPIAMNCSNL WGNFSYGSIC SFHCLEGQLL NGSAQTACQE NGHWSTTVPT CQAGPLTIQE ALTYFGGAVA STIGLIMGGT LLALLRKRFR QKDDGKCPLN PHSHLGTYGV FTNAAFDPSP | |
Greater than 95% as determined by SDS-PAGE. Single band on Western Blot. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
P-Selectin is supplied in 50% Glycerol. |