missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Osteoprotegerin / TNFRSF11B Protein

A DNA sequence encoding the Human Osteoprotegerin / TNFRSF11B, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10427-ec-50ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983818

  • 313.00 CHF / 50 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_002537.3
Osteoprotegerin / TNFRSF11B Protein
50 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ETFPPKYLHY DEETSHQLLC DKCPPGTYLK QHCTAKWKTV CAPCPDHYYT DSWHTSDECL YCSPVCKELQ YVKQECNRTH NRVCECKEGR YLEIEFCLKH RSCPPGFGVV QAGTPERNTV CKRCPDGFFS NETSSKAPCR KHTNCSVFGL LLTQKGNATH DNICSGNSES TQK
4982
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
TNFRSF11B
The ED50 as determined by neutralizing the stimulation of U937 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg in the presence of 10 ng/mL soluble rHuRANKL (sRANKL).
Human
Untagged
0.2 um filtered 30 % acetonitrile, 0.1 % TFA, lyophilized
Videos
Dokumentation