missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OSR2 Protein
A cDNA sequence encoding the OSR2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12934-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
OSR2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGSKALP APIPLHPSLQ LTNYSFLQAV NTFPATVDHL QGLYGLSAVQ TMHMNHWTLG YPNVHEITRS TITEMAAAQG LVDARFPFPA LPFTTHLFHP KQGAIAHVLP ALHKDRPRFD FANLAVAATQ EDPPKMGDLS KLSPGLGSPI SGLSKLTPDR KPSRGRLPSK TKKEFICKFC GRHFTKSYNL LIHERTHTDE RPYTCDICHK AFRRQDHLRD HRYIHSKEKP FKCQECGKGF CQSRTLAVHK TLHMQTSSPT AASSAAKCSG ETVICGGTA | |
Greater than 80% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
OSR2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |