missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ornithine Aminotransferase Protein
A cDNA sequence encoding the Ornithine Aminotransferase was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12932-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Ornithine Aminotransferase Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTVQGPPTSD DIFEREYKYG AHNYHPLPVA LERGKGIYLW DVEGRKYFDF LSSYSAVNQG HCHPKIVNAL KSQVDKLTLT SRAFYNNVLG EYEEYITKLF NYHKVLPMNT GVEAGETACK LARKWGYTVK GIQKYKAKIV AAGNFWGRT LSAISSSTDP TSYDGFGPFM PGFDIIPYND LPALERALQD PNVAAFMVEP IQGEAGVVVP DPGYLMGVRE LCTRHQVLFI ADEIQTGLAR TGRWLAVDYE NVRPDIVLLG KALSGGLYPV SAVLCDDDIM LTIKPGEHGS TYGGNPLGCR VAIAALEVLE EENLAENADK LGIILRNELM KLPSDVVTAV RGKGLLNAIV IKETKDWDAW KVCLRLRDNG LLAKPTHGDI IRFAPPLVIK EDELRESIEI INKTILSF | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Ornithine Aminotransferase protein solution contains 20mM Tris, pH 8.0, and 10% glycerol. |