missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ORAOV1 Protein

A cDNA sequence encoding the ORAOV1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12930-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911989

  • 188.00 CHF / 10 Mikrogramm


Verfügbar ab
26-03-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

ORAOV1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ORAOV1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAGSQD IFDAIVMADE RFHGEGYREG YEEGSSLGVM EGRQHGTLHG AKIGSEIGCY QGFAFAWKCL LHSCTTEKDS RKMKVLESLI GMIQKFPYDD PTYDKLHEDL DKIRGKFKQF CSLLNVQPDF KISAEGSGLS F
Videos
Dokumentation