missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT14 Protein
A cDNA sequence encoding the NUDT14 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12901-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NUDT14 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMERIEGA SVGRCAASPY LRPLTLHYRQ NGAQKSWDFM KTHDSVTVLL FNSSRRSLVL VKQFRPAVYA GEVERRFPGS LAAVDQDGPR ELQPALPGSA GVTVELCAGL VDQPGLSLEE VACKEAWEEC GYHLAPSDLR RVATYWSGVG LTGSRQTMFY TEVTDAQRSG PGGGLVEEGE LIEVVHLPLE GAQAFADDPD IPKTLGVIFG VSWFLSQVAP NLDLQ | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NUDT14 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |