missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRG1 Protein
A DNA sequence encoding the Human NRG1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10414-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001153467.1 | |
NRG1 Protein | |
Research Use Only | |
NRG1 | |
The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 m filtered 1 M PBS, pH 7.4, with 5 % trehalose, lyophilized |
3084 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE | |
Greater than 97% as determined by SDS-PAGE and HPLC. |