missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NOP16 Protein
A cDNA sequence encoding the NOP16 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12864-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
51491 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKAKGK TRRQKFGYSV NRKRLNRNAR RKAAPRIECS HIRHAWDHAK SVRQNLAEMG LAVDPNRAVP LRKRKVKAME VDIEERPKEL VRKPYVLNDL EAEASLPEKK GNTLSRDLID YVRYMVENHG EDYKAMARDE KNYYQDTPKQ IRSKINVYKR FYPAEWQDFL DSLQKRKMEV E | |
Greater than 85.0% as determined by SDS-PAGE. |
NOP16 Protein | |
Research Use Only | |
NOP16 | |
Human | |
His | |
NOP16 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM EDTA and 2mM DTT. |