missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMRAL1 Protein
A cDNA sequence encoding the NMRAL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12859-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NMRAL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMVDKKL VVVFGGTGAQ GGSVARTLLE DGTFKVRVVT RNPRKKAAKE LRLQGAEVVQ GDQDDQVIME LALNGAYATF IVTNYWESCS QEQEVKQGKL LADLARRLGL HYVVYSGLEN IKKLTAGRLA AAHFDGKGEV EEYFRDIGVP MTSVRLPCYF ENLLSHFLPQ KAPDGKSYLL SLPTGDVPMD GMSVSDLGPV VLSLLKMPEK YVGQNIGLST CRHTAEEYAA LLTKHTRKVV HDAKMTPEDY EKLGFPGARD LANMFRFYAL RPDRDIELTL RLNPKALTLD QWLEQHKGDF NLL | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NMRAL1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 40% glycerol. |