missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NIPSNAP1 Protein
A cDNA sequence encoding the NIPSNAP1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12846-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NIPSNAP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAPRLCS ISVTARRLLG GPGPRAGDVA SAAAARFYSK DNEGSWFRSL FVHKVDPRKD AHSTLLSKKE TSNLYKIQFH NVKPEYLDAY NSLTEAVLPK LHLDEDYPCS LVGNWNTWYG EQDQAVHLWR FSGGYPALMD CMNKLKNNKE YLEFRRERSQMLLSRRNQLL LEFSFWNEPQ PRMGPNIYEL RTYKLKPGTM IEWGNNWARA IKYRQENQEA VGGFFSQIGE LYVVHHLWAY KDLQSREETR NAAWRKRGWD ENVYYTVPLV RHMESRIMIP LKISPLQ | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NIPSNAP1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |