missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NHLH2 Protein
A cDNA sequence encoding the NHLH2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12844-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NHLH2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMLSPDQ AADSDHPSSA HSDPESLGGT DTKVLGSVSD LEPVEEAEGD GKGGSRAALY PHPQQLSREE KRRRRRATAK YRSAHATRER IRVEAFNLAF AELRKLLPTL PPDKKLSKIE ILRLAICYIS YLNHVLDV | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NHLH2 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.2M NaCl, 50% glycerol and 5mM DTT. |