missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NHLH2 Protein
A cDNA sequence encoding the NHLH2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | NHLH2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
15921729
|
enQuireBio™
QP12844-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15901729
|
enQuireBio™
QP12844-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15911729
|
enQuireBio™
QP12844-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Specifications
NHLH2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NHLH2 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.2M NaCl, 50% glycerol and 5mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMLSPDQ AADSDHPSSA HSDPESLGGT DTKVLGSVSD LEPVEEAEGD GKGGSRAALY PHPQQLSREE KRRRRRATAK YRSAHATRER IRVEAFNLAF AELRKLLPTL PPDKKLSKIE ILRLAICYIS YLNHVLDV | |
Greater than 90.0% as determined by SDS-PAGE. |