missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFB9 Protein

A cDNA sequence encoding the NDUFB9 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12817-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911649

  • 125.00 CHF / 2 Mikrogramm


Verfügbar ab
23-03-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

4715
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
NDUFB9
Human
His
The NDUFB9 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
NDUFB9 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAFLASG PYLTHQQKVL RLYKRALRHL ESWCVQRDKY RYFACLMRAR FEEHKNEKDM AKATQLLKEA EEEFWYRQHP QPYIFPDSPG GTSYERYDCY KVPEWCLDDW HPSEKAMYPD YFAKREQWKK LRRESWEREV KQLQEETPPG GPLTEALPPA RKEGDLPPLW WYIVTRPRER PM
Videos
Dokumentation