missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAIF1 Protein
A cDNA sequence encoding the NAIF1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Gen-ID (Entrez) | 203245 |
---|---|
Name | NAIF1 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | NAIF1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15931539
|
enQuireBio™
QP12786-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15921539
|
enQuireBio™
QP12786-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15911539
|
enQuireBio™
QP12786-100UG |
100 μg |
1735.00 CHF
100 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
203245 | |
Research Use Only | |
NAIF1 | |
Human | |
His | |
NAIF1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |
NAIF1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAVPAKK RKMNFSEREV EIIVEELELK KHLLVNHFNA GVPLAAKSAA WHGILRRVNA VATCRRELPE VKKKWSDLKT EVRRKVAQVR AAVEGGEAPG PTEEDGAGGP GTGGGSGGGG PAVAPVLLTP MQQRICNLLG EATIISLPST TEIHPVALGP SATAAAATVT LTQIPTETTY HTLEEGVVEY CTAEAPPPLP PETPVDMMAQ HADTSVKPQA LKSRIALNSA KLIQEQRVTN LHVKEIAQHL EQQNDLLQMI RRSQEVQACA QERQAQAMEG TQAALSVLIQ VLRPMIKDFR RYLQSNTANP APASDPGQVA QNGQPDSIIQ | |
Greater than 90.0% as determined by SDS-PAGE. |