missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NACA Protein
A cDNA sequence encoding the NACA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12782-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NACA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPGEATETVP ATEQELPQPQ AETGSGTESD SDESVPELEE QDSTQATTQQ AQLAAAAEID EEPVSKAKQS RSEKKARKAM SKLGLRQVTG VTRVTIRKSK NILFVITKPD VYKSPASDTY IVFGEAKIED LSQQAQLAAA EKFKVQGEAV SNIQENTQTP TVQEESEEEE VDETGVEVKD IELVMSQANV SRAKAVRALK NNSNDIVNAI MELTM | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NACA solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |