Learn More
enQuireBio™ Recombinant Human MYOZ1 Protein
Click to view available options
Menge:
1 mg
20 μg
5 μg
Packungsgröße:
1 Milligramm
20 Mikrogramm
5 Mikrogramm
Spezifikation
Spezifikation
| Name | MYOZ1 Protein |
| Menge | 5 μg |
| Kennzeichnung | Research Use Only |
| Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Produkttyp | Recombinant Protein |
| Kreuzreaktivität | Human |
| Spezies | E. coli |
| Proteinmarkierung | His |
| Sequenz | MGSSHHHHHH SSGLVPRGSH MGSMPLSGTP APNKKRKSSK LIMELTGGGQ ESSGLNLGKK ISVPRDVMLE ELSLLTNRGS KMFKLRQMRV EKFIYENHPD VFSDSSMDHF QKFLPTVGGQ LGTAGQGFSY SKSNGRGGSQ AGGSGSAGQY GSDQQHHLGS GSGAGGTGGP AGQAGRGGAA GTAGVGETGS GDQAGGEGKH ITVFKTYISP WERAMGVDPQ QKMELGIDLL AYGAKAELPK YKSFNRTAMP YGGYEKASKR MTFQMPKFDL GPLLSEPLVL YNQNLSNRPS FNRTPIPWLS SGEPVDYNVD IGIPLDGETE EL |
| Puffer | MYOZ1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M UREA and 10% glycerol. |
| Mehr anzeigen |