missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYL12B Protein
A cDNA sequence encoding the MYL12B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12768-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
MYL12B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSSKKA KTKTTKKRPQ RATSNVFAMF DQSQIQEFKE AFNMIDQNRD GFIDKEDLHD MLASLGKNPT DAYLDAMMNE APGPINFTMF LTMFGEKLNG TDPEDVIRNA FACFDEEATG TIQEDYLREL LTTMGDRFTD EEVDELYREA PIDKKGNFNY IEFTRILKHG AKDKDD | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MYL12B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM Nacl, 2mM DTT and 40% glycerol. |