missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MSRB2 Protein
A cDNA sequence encoding the MSRB2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12744-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
MSRB2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVRGQAGGGG PGTGPGLGEA GSLATCELPL AKSEWQKKLT PEQFYVTREK GTEPPFSGIY LNNKEAGMYH CVCCDSPLFS SEKKYCSGTG WPSFSEAHGT SGSDESHTGI LRRLDTSLGS ARTEVVCKQC EAHLGHVFPD GPGPNGQRFC INSVALKFKP RKH | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MSRB2 protein solution (1 mg/1ml) is formulated in In 20 mM Tris-HCl Buffer (pH 7.5), 1 mM DTT, 0.1 mM PMSF and 10% Glycerol. |