missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS28 Protein
A cDNA sequence encoding the MRPS28 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | MRPS28 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15921379
|
enQuireBio™
QP12735-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15911379
|
enQuireBio™
QP12735-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15901379
|
enQuireBio™
QP12735-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
MRPS28 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPS28 protein solution (0.25 mg/ml) containing 20mM Phosphate buffer saline (pH 8.0), 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGSPKNVE SFASMLRHSP LTQMGPAKDK LVIGRIFHIV ENDLYIDFGG KFHCVCRRPE VDGEKYQKGT RVRLRLLDLE LTSRFLGATT DTTVLEANAV LLGIQESKDS RSKEEHHEK | |
Greater than 90.0% as determined by SDS-PAGE. |